Order Entry
Czech Republic
ContactUsLinkComponent
1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole 95%
1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole 95%
Catalog # APOSOR32025-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Phenyl-1H-pyrazol-4-yl)-1H-1,2,4-triazole 95%
Catalog # APOSOR32025-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.223 g/mol
Storage Temperature:Ambient
CAS Number:241146-80-5
Formula:C₁₁H₉N₅
MDL Number:MFCD00214691