{Hexahydro-2H-[1,2]oxazolo[2,3-a]pyridin-2-yl}methanol 95%
Catalog # APOSOR32019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:157.21 g/mol
Storage Temperature:Ambient
CAS Number:241132-72-9
Formula:C₈H₁₅NO₂
MDL Number:MFCD00140539