Order Entry
Czech Republic
ContactUsLinkComponent
N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide 95%
N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide 95%
Catalog # APOSOR32008-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Ethyl-5-(1-phenoxyethyl)-1H-pyrazole-1-carboxamide 95%
Catalog # APOSOR32008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.304 g/mol
Storage Temperature:Ambient
CAS Number:241127-12-8
Formula:C₁₄H₁₇N₃O₂
MDL Number:MFCD00139095