Order Entry
Czech Republic
ContactUsLinkComponent
 
4-[4-(Pyridin-2-yl)-1H-pyrazol-3-yl]morpholine 95%
Catalog # APOSOR32001-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[4-(Pyridin-2-yl)-1H-pyrazol-3-yl]morpholine 95%
Catalog # APOSOR32001-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.266 g/mol
Storage Temperature:Ambient
CAS Number:240115-91-7
Formula:C₁₂H₁₄N₄O
MDL Number:MFCD00127011