Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl cis-3-hydroxycyclobutanecarboxylate
Catalog # APOSOR317100-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl cis-3-hydroxycyclobutanecarboxylate
Catalog # APOSOR317100-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :130.142 g/mol
Storage Temperature:Ambient
CAS Number:63485-50-7
Formula:C₆H₁₀O₃
MDL Number:MFCD18483124