
Methyl cis-3-(Boc-amino)cyclobutanecarboxylate
Catalog # APOSOR317090-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.273 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:142733-63-9
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD20922913