Methyl cis-3-Amino-cyclobutanecarboxylate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1212304-86-3
:
Formula:C₆H₁₁NO₂·ClH
MDL Number:MFCD11099895
:165.618 g/mol
Storage Temperature:Ambient
Frequently Bought Together