Order Entry
Czech Republic
ContactUsLinkComponent
1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride 95%
1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride 95%
Catalog # APOSOR315878-25MG
Supplier:  Apollo Scientific
CAS Number:  
1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride 95%
Catalog # APOSOR315878-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :219.712 g/mol
Storage Temperature:Ambient
CAS Number:2208138-91-2
Formula:C₉H₁₇N₃O·ClH
MDL Number:MFCD30489690