
2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester 97%
Catalog # APOSOR315428-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.258 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:152533-47-6
Formula:C₁₁H₁₇NO₃
MDL Number:MFCD18633018