
Spiro[isoindole-1,4'-piperidin]-3(2H)-one hydrochloride
Catalog # APOSOR313019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.713 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:788812-21-5
Formula:C₁₂H₁₄N₂O·ClH
MDL Number:MFCD18711590