Order Entry
Czech Republic
ContactUsLinkComponent
(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol
(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol
Catalog # APOSOR311294-500MG
Supplier:  Apollo Scientific
CAS Number:  
(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol
Catalog # APOSOR311294-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :214.689 g/mol
Storage Temperature:Ambient
CAS Number:139406-15-8
Formula:C₁₁H₁₅ClO₂
MDL Number:MFCD28976644