3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-1-ol
Catalog # APOSOR311253-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.152 g/mol
Storage Temperature:Refrigerator
CAS Number:651030-57-8
Formula:C₁₅H₂₃BO₃
MDL Number:MFCD18734359