Order Entry
Czech Republic
ContactUsLinkComponent
5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate
5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate
Catalog # APOSOR310958-500MG
Supplier:  Apollo Scientific
CAS Number:  
5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate
Catalog # APOSOR310958-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :295.334 g/mol
Storage Temperature:Ambient
CAS Number:1286753-88-5
Formula:C₁₄H₂₁N₃O₄
MDL Number:MFCD18837793