Methyl 3-(4-methoxyphenyl)-3-methylbutanoate
Catalog # APOSOR310919-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:222.28 g/mol
Storage Temperature:Ambient
CAS Number:67437-37-0
Formula:C₁₃H₁₈O₃
MDL Number:MFCD22688506