
5,5-Dimethyl-hexahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-5'-one
Catalog # APOSOR310550-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.296 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:92007-37-9
Formula:C₁₃H₂₀O₃
MDL Number:MFCD27957346