4-[(Biphenyl-2-ylcarbonyl)amino]-2-chlorobenzoic acid
Catalog # APOSOR310539-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:351.783 g/mol
Storage Temperature:Ambient
CAS Number:180340-01-6
Formula:C₂₀H₁₄ClNO₃
MDL Number:MFCD20266812