(1S)-2,2-Dimethyl-1-phenylpropane-1,3-diol
Catalog # APOSOR310533-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:180.244 g/mol
Storage Temperature:Ambient
CAS Number:133164-39-3
Formula:C₁₁H₁₆O₂
MDL Number:MFCD24444156