Order Entry
Czech Republic
ContactUsLinkComponent
5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]
5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]
Catalog # APOSOR310531-1G
Supplier:  Apollo Scientific
CAS Number:  
5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]
Catalog # APOSOR310531-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.297 g/mol
Storage Temperature:Ambient
CAS Number:92007-41-5
Formula:C₁₃H₂₀O₂
MDL Number:MFCD27957338