Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate
Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate
Catalog # APOSOR310526-500MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate
Catalog # APOSOR310526-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :282.332 g/mol
Storage Temperature:Ambient
CAS Number:105448-66-6
Formula:C₁₅H₂₂O₅
MDL Number:MFCD27957337