Order Entry
Czech Republic
ContactUsLinkComponent
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
Catalog # APOSOR310472-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
Catalog # APOSOR310472-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.093 g/mol
Storage Temperature:Ambient
CAS Number:862081-37-6
Formula:C₁₄H₁₇BO₄
MDL Number:MFCD18251723