Order Entry
Czech Republic
ContactUsLinkComponent
1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
Catalog # APOSOR310428-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
Catalog # APOSOR310428-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :302.795 g/mol
Storage Temperature:Ambient
CAS Number:103628-22-4
Formula:C₁₈H₁₉ClO₂
MDL Number:MFCD09835550