Methyl 3-(benzyloxy)-5-hydroxybenzoate
Catalog # APOSOR310423-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:258.269 g/mol
Storage Temperature:Ambient
CAS Number:54915-31-0
Formula:C₁₅H₁₄O₄
MDL Number:MFCD06655985