Order Entry
Czech Republic
ContactUsLinkComponent
Methyl (1R,8S,9S,Z)-bicyclo[6.1.0]non-4-ene-9-carboxylate
Methyl (1R,8S,9S,Z)-bicyclo[6.1.0]non-4-ene-9-carboxylate
Catalog # APOSOR310408-500MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl (1R,8S,9S,Z)-bicyclo[6.1.0]non-4-ene-9-carboxylate
Catalog # APOSOR310408-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :180.244 g/mol
Storage Temperature:Ambient
CAS Number:61452-51-5
Formula:C₁₁H₁₆O₂
MDL Number:MFCD27924238