Zadání objednávky
Czech Republic
ContactUsLinkComponent
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid
Irritant
Health
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid
Catalog # APOSOR310381-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Health
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid
Catalog # APOSOR310381-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :393.475 g/mol
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1058705-57-9
Formula:C₂₄H₂₇NO₄
MDL Number:MFCD03094931