5-Bromo-3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:875639-78-4
:
Formula:C₁₄H₁₁BrN₂O
MDL Number:MFCD26523310
:303.154 g/mol
Storage Temperature:Ambient
Frequently Bought Together