4-(Hydroxymethyl)-4-phenylcyclohexan-1-one
Catalog # APOSOR310320-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.265 g/mol
Storage Temperature:Ambient
CAS Number:51510-01-1
Formula:C₁₃H₁₆O₂
MDL Number:MFCD17013064