Order Entry
Czech Republic
ContactUsLinkComponent
(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
Catalog # APOSOR310083-500MG
Supplier:  Apollo Scientific
CAS Number:  
(2S)-2-[(3aR,4R,6R,7aS)-3a,5,5-Trimethylhexahydro-4,6-methano-1,2-benzoxaborol-2(3H)-yl]pyrrolidine hydrochloride
Catalog # APOSOR310083-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :283.645 g/mol
Storage Temperature:Ambient
CAS Number:149716-72-3
Formula:C₁₅H₂₆BNO·ClH
MDL Number:MFCD21602036