(2-Bromo-5-methoxy-4-methylphenyl)methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57295-33-7
:
Formula:C₉H₁₁BrO₂
MDL Number:MFCD21605749
:231.086 g/mol
Melting Pt:97…99 °C
Storage Temperature:Ambient
Frequently Bought Together