Order Entry
Czech Republic
ContactUsLinkComponent
(1R,2R,3R,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid
(1R,2R,3R,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid
Catalog # APOSOR309443-100MG
Supplier:  Apollo Scientific
CAS Number:  
(1R,2R,3R,5R)-(2-tert-Butoxycarbonylamino)-6,6-dimethylbicyclo[3.1.1]heptan-3-carboxylic acid
Catalog # APOSOR309443-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :283.363 g/mol
Storage Temperature:Ambient
CAS Number:1027343-75-4
Formula:C₁₅H₂₅NO₄
MDL Number:MFCD09881422