Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate
tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate
Catalog # APOSOR309438-100MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl (1R,2R,3S,5R)-(2-methylaminocarbonyl-6,6-dimethylbicyclo[3.1.1]hept-3-yl)-carbamate
Catalog # APOSOR309438-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :296.405 g/mol
Storage Temperature:Ambient
CAS Number:1027343-56-1
Formula:C₁₆H₂₈N₂O₃
MDL Number:MFCD09881417