Ethyl 4-chloro-6,7-dimethoxyquinazoline-2-carboxylate
Catalog # APOSOR309400-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:296.706 g/mol
Storage Temperature:Ambient
CAS Number:1189105-82-5
Formula:C₁₃H₁₃ClN₂O₄
MDL Number:MFCD12675151