4-Chloro-6,8-dimethyl-2-phenylquinoline
Catalog # APOSOR309307-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.753 g/mol
Storage Temperature:Ambient
CAS Number:101602-31-7
Formula:C₁₇H₁₄ClN
MDL Number:MFCD12674825