Order Entry
Czech Republic
ContactUsLinkComponent
4,8-Dibromo-2-methylquinoline
4,8-Dibromo-2-methylquinoline
Catalog # APOSOR309293-1G
Supplier:  Apollo Scientific
CAS Number:  
4,8-Dibromo-2-methylquinoline
Catalog # APOSOR309293-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :300.977 g/mol
Storage Temperature:Ambient
CAS Number:1070879-55-8
Formula:C₁₀H₇Br₂N
MDL Number:MFCD11505139