4,6-Dibromo-2-methylquinoline
Catalog # APOSOR309292-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:300.977 g/mol
Storage Temperature:Ambient
CAS Number:1070879-53-6
Formula:C₁₀H₇Br₂N
MDL Number:MFCD09261090