Order Entry
Czech Republic
ContactUsLinkComponent
6-Ethyl-4-hydroxyquinoline
6-Ethyl-4-hydroxyquinoline
Catalog # APOSOR309214-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Ethyl-4-hydroxyquinoline
Catalog # APOSOR309214-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.211 g/mol
Storage Temperature:Ambient
CAS Number:303121-13-3
Formula:C₁₁H₁₁NO
MDL Number:MFCD02332229