6-Chloro-8-methylquinoline-3-carboxylic acid
Catalog # APOSOR309211-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.64 g/mol
Storage Temperature:Ambient
CAS Number:948289-56-3
Formula:C₁₁H₈ClNO₂
MDL Number:MFCD09787823