Order Entry
Czech Republic
ContactUsLinkComponent
6-Bromo-2-methylquinoline-3-carboxylic acid ≥98%
Irritant
6-Bromo-2-methylquinoline-3-carboxylic acid ≥98%
Catalog # APOSOR309203-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-2-methylquinoline-3-carboxylic acid ≥98%
Catalog # APOSOR309203-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :266.091 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:92513-39-8
Formula:C₁₁H₈BrNO₂
MDL Number:MFCD09787811