Order Entry
Czech Republic
ContactUsLinkComponent
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 97%
Irritant
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 97%
Catalog # APOSOR309123-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 97%
Catalog # APOSOR309123-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.3 g/mol
Melting Pt:74…77 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:1245816-29-8
Formula:C₁₂H₂₁NO₃
MDL Number:MFCD12545892