
2-(Piperidin-1-yl)-1,3-thiazole-5-carboxylic acid
Catalog # APOSOR30810-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:212.269 g/mol
Melting Pt:146…151 °C
:GHS07
Storage Temperature:Ambient
CAS Number:180403-13-8
Formula:C₉H₁₂N₂O₂S
MDL Number:MFCD11506352