
1-tert-Butyl 2-methyl (2R)-piperazine-1,2-dicarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:252990-05-9
:
Formula:C₁₁H₂₀N₂O₄
MDL Number:MFCD04115327
:244.287 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together