Order Entry
Czech Republic
ContactUsLinkComponent
6,7-Dimethylquinoline-3-carboxylic acid
6,7-Dimethylquinoline-3-carboxylic acid
Catalog # APOSOR307998-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dimethylquinoline-3-carboxylic acid
Catalog # APOSOR307998-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.221 g/mol
Storage Temperature:Ambient
CAS Number:948294-50-6
Formula:C₁₂H₁₁NO₂
MDL Number:MFCD09787801