Order Entry
Czech Republic
ContactUsLinkComponent
4,6-Dichloro-2-trichloromethyl-quinoline
4,6-Dichloro-2-trichloromethyl-quinoline
Catalog # APOSOR307815-5G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dichloro-2-trichloromethyl-quinoline
Catalog # APOSOR307815-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :315.41 g/mol
Storage Temperature:Ambient
CAS Number:93600-20-5
Formula:C₁₀H₄Cl₅N
MDL Number:MFCD00272424