
(1R,2R)-4-Oxo-cyclopentane-1,2-dicarboxylic acid dimethyl ester
Catalog # APOSOR307802-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:200.189 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:35079-19-7
Formula:C₉H₁₂O₅
MDL Number:MFCD10001388