
5,7-Dihydroxy-2,2-dimethylchroman-4-one 95%
Catalog # APOSOR307797-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.211 g/mol
Melting Pt:187…193 °C
:GHS07
Storage Temperature:Ambient
CAS Number:883-09-0
Formula:C₁₁H₁₂O₄
MDL Number:MFCD02066217