4-(Pyrrolidin-1-ylmethyl)aniline hydrochloride
Catalog # APOSOR307768-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.719 g/mol
Storage Temperature:Ambient
CAS Number:866956-98-1
Formula:C₁₁H₁₆N₂·ClH
MDL Number:MFCD08447180