1-(3,4-Dimethoxyphenyl)cyclopropanecarbonitrile
Catalog # APOSOR307752-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.237 g/mol
Storage Temperature:Ambient
CAS Number:20802-15-7
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD01314306