3-Benzo[1,3]dioxol-5-ylmethyl-piperidine
Catalog # APOSOR307735-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.28 g/mol
Storage Temperature:Ambient
CAS Number:955314-95-1
Formula:C₁₃H₁₇NO₂
MDL Number:MFCD05863607