Order Entry
Czech Republic
ContactUsLinkComponent
N-Hydroxy-2,3:5,6-bis-O-(1-methylethylidene)-α-D-glucofuranosylamine
N-Hydroxy-2,3:5,6-bis-O-(1-methylethylidene)-α-D-glucofuranosylamine
Catalog # APOSOR307695-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Hydroxy-2,3:5,6-bis-O-(1-methylethylidene)-α-D-glucofuranosylamine
Catalog # APOSOR307695-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.298 g/mol
Storage Temperature:Ambient
CAS Number:1226812-52-7
Formula:C₁₂H₂₁NO₆
MDL Number:MFCD15530194