
(1R*,2S*,4S*)-7-Aza-bicyclo[2.2.1]heptane-2,7-dicarboxylic acid 7-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:500556-91-2
:
Formula:C₁₂H₁₉NO₄
MDL Number:MFCD12198742
:241.284 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together