Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
APOSOR307670-1G
:
Apollo Scientific
:
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
APOSOR307670-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
227.302 g/mol
:
Ambient
:
20616-52-8
:
C₁₅H₁₇NO
:
MFCD11505079